Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB5384A4

Sigma-Aldrich

Anti-NG2 Antibody, clone 132.39, Alexa Fluor488 Conjugate

clone 132.39, from mouse, ALEXA FLUOR 488

Synonym(s):

Chondroitin sulfate proteoglycan 4, Chondroitin sulfate proteoglycan NG2, HSN tumor-specific antigen

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$699.00

$699.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$699.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$699.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

132.39, monoclonal

species reactivity

rat

species reactivity (predicted by homology)

mouse, human

technique(s)

immunocytochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CSPG4(1464)
mouse ... Cspg4(121021)
rat ... Cspg4(81651)

General description

NG2 is a high molecular weight, integral membrane chondroitin sulfate proteoglycan. It is found on the surfaces of an unusual class of glial cells within the developing and mature central nervous system that have the properties of oligodendrocyte precursor cells (i.e., O-2A progenitor cells). NG2 is also found on the surfaces of chondroblasts, proliferating capillary endothelial cells, some human melanoma cell lines, and on leukemic blasts in childhood acute lymphoblastic leukemia. The NG2 proteoglycan is likely to play a role in regulation of cell motility, axon outgrowth and the cellular responses to platelet-derived growth factor.

Specificity

This antibody recognizes the native, recombinant, and protein from cells which express NG2.

Immunogen

Cell line expressing a truncated form of NG2

Application

Anti-NG2 Antibody, clone 132.39, Alexa Fluor488 Conjugate detects level of NG2 & has been published & validated for use in ICC.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Immunocytochemistry in rat oligodendrocyte precursor cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected NG2 in rat oligodendrocyte precursor cells. Performed without Triton.

Target description

The unconjugated parent antibody (Cat. No. MAB5384) has an observed molecular weight at ~280 kDa

Physical form

Protein G Purified
Purified mouse monoclonal IgG1 conjugated to Alexa Fluor 488 in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Rat oligodendrocyte precursor cells

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annika Andersson-Sjöland et al.
Laboratory investigation; a journal of technical methods and pathology, 96(2), 206-217 (2015-09-15)
Pulmonary fibrosis is a grave diagnosis with insidious progression, generally considered as a consequence of aberrant epithelial wound healing and excessive scarring. This process is commonly modeled in animals by local bleomycin administration, resulting in peribronchial inflammation and subsequent fibrosis.
Yitao Zhu et al.
Nature (2024-08-31)
Human mutations in neuropeptide Y (NPY) have been linked to high body mass index but not altered dietary patterns1. Here we uncover the mechanism by which NPY in sympathetic neurons2,3 protects from obesity. Imaging of cleared mouse brown and white adipose
Kristina Rydell-Törmänen et al.
Scientific reports, 4, 6383-6383 (2014-09-19)
The ECM exerts great effects on cells, and changed composition may therefore have profound impact. Small leucine-rich proteoglycans, e.g. fibromodulin, are essential in collagen assembly. Our aim was to investigate the role of fibromodulin in healthy and fibrotic lung parenchyma

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service