Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB5290

Sigma-Aldrich

Anti-Dopamine D1A Receptor Antibody, clone SG2-D1a

clone SG2-D1a, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SG2-D1a, monoclonal

species reactivity

mouse, sheep, rat

should not react with

pig, rabbit, human, bovine, canine

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DRD1(1812)

Specificity

Dopamine Receptor D1A, no cross reactivity to D1B (D5) receptor. Immunoreactivity was found to be specific for mouse and rat D1A receptor. Human, pig, dog, rabbit, sheep and bovine samples were non-reactive by western analysis.

Immunogen

Recombinant rat D1a Receptor, carboxy terminus.

Application

Anti-Dopamine D1A Receptor Antibody, clone SG2-D1a detects level of Dopamine D1A Receptor & has been published & validated for use in IH & WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot: 1:250-1:500 on unboiled membrane fractions from rat caudate tissue. Reactivity is also seen in perfrontal and temporal cortex. An identified band between 45-66kDa is observed (Luedtke, 1999).

Immunohistochemistry: 4% PFA fixed, vibratome sections, blocked with 3% NGS, 1% BSA with 0.2% triton X-100. Primary antibodies diluted onlyin PBS with excess blocking reagents.

Optimal working dilutions must be determined by the end user.

SUGGESTED WESTERN BLOT PROTOCOL

Solutions

10X Transfer buffer 1 liter

Tris 115.3 gm

Glycine 24.2 gm

[SDS 4 gm (may be omitted)]

For a 1X working solution: 1 liter

mix 700 mL water

200 mL MeOH

100 mL 10X TransferTBST

stock 1 liter

1M Tris pH 7.5 10 mL

5 M NaC 30 mL

10% Tween-20 5 mL

1. Run SDS-PAGE gel as desired.

2. Soak PVDF membrane (ImmobilonP from Millipore) in 100% methanol for 1-2 minutes to wet the membrane.

3. Soak membrane in transfer buffer (see below) until ready to set up blot transfer (>15 min).

4. Assemble gel-transfer sandwich with the blot toward the positive electrode.

5. Transfer protein from gel to membrane at 100 volts for 1-2 hours for small molecular weight proteins or 65 mA for 16 hours (overnight) for complete transfer of higher molecular weight proteins.

6. Stain the transferred bands with Chemicon BLOT-FastStain (Catalog Number 2076).

7. Destain with deionized water.

8. Block with 5% non-fat milk (Marvel or Carnation) in TBS or PBS, overnight at 4°C. The non-fat milk should be dissolved freshly, centrifuged 10,000 rpm for 10 min, and filtered through glass filter (Gelman Acrodisc).

9. Incubation with first antibody overnight with gentle agitation at 4°C.

Optimal working dilutions and incubation time will need to be determined by the end user.

10. Wash at least 3 x 5 min. with TBST. From this stage, azide should be omitted.

11. Incubation with the secondary antibody (HRP-conjugated goat anti-mouse antibody, for example Chemicon Catalog Number AP124P, diluted appropriately) 1 h at room temperature.

12. Wash at least 3 x 5 min. with TBST.

13. Perform ECL with commercial kit (femtoLUCENT, Chemicon Catalog Number 2078).

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02 M Phosphate buffer, 0.25 M NaCl, pH 7.6 with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlo Cavallotti et al.
Neurochemical research, 29(8), 1499-1504 (2004-07-21)
Dopamine receptors (Dar) were studied as a component of the nervous dopaminergic system in the human dura mater. Dar were stained in several dural zones (vascular, perivascular, intervascular) in different regions (basal, calvarial, tentorial, occipital, frontal, parietal, temporal) of the
Laura E O'Dell et al.
Addiction biology, 19(6), 1006-1019 (2013-07-10)
Patients with diabetes display a heightened propensity to use tobacco; however, it is unclear whether they experience enhanced rewarding effects of nicotine. Thus, this study examined the reinforcing effects of nicotine in a rodent model of diabetes involving administration of
Chenhao Li et al.
International journal of molecular sciences, 24(23) (2023-12-09)
Clinical imaging studies have revealed that the hypothalamus is activated in migraine patients prior to the onset of and during headache and have also shown that the hypothalamus has increased functional connectivity with the spinal trigeminal nucleus. The dopaminergic system
Anouk Schrantee et al.
PloS one, 12(2), e0172776-e0172776 (2017-02-28)
Dexamphetamine (AMPH) is a psychostimulant drug that is used both recreationally and as medication for attention deficit hyperactivity disorder. Preclinical studies have demonstrated that repeated exposure to AMPH can induce damage to nerve terminals of dopamine (DA) neurons. We here
Mizuki Maeda et al.
The Journal of veterinary medical science, 83(3), 542-548 (2021-02-02)
Recently, it has been reported that neonicotinoid pesticides (NNs) are transferred from mother to child and are assumed to affect the next generation, but the behavioral effects of NN exposure at different developmental stages have not been investigated. We exposed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service