Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB4617

Sigma-Aldrich

Anti-Survivin Antibody, clone 8E2

clone 8E2, Chemicon®, from mouse

Synonym(s):

TIAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8E2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable

isotype

IgG1

suitability

not suitable for Western blot

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BIRC5(332)

General description

Survivin, an inhibitor of apoptosis (IAP) protein, is expressed in the G2/M phase of the cell cycle in a cycle-regulated manner. At the beginning of mitosis, survivin associates with microtubules of the mitotic spindle in a specific and saturable reaction that is regulated by microtubule dynamics. Disruption of survivin-microtubule interactions results in loss of survivin′s anti-apoptosis function and increased caspase-3 activity, a mechanism involved in cell death, during mitosis. It indicates that survivin may counteract a default induction of apoptosis in G2/M phase. The overexpression of survivin in cancer may overcome this apoptotic checkpoint and favor aberrant progression of transformed cells through mitosis.

Specificity

MAB4617 detects human survivin (16.5 kDa).

Immunogen

Full length recombinant human survivin

Application

Detect Survivin using this Anti-Survivin Antibody, clone 8E2 validated for use in ELISA & IF.
Immunofluorescence

ELISA (for detection only)

Does not work for Western Blot

Does not work for Immunohistochemistry

Optimal working dilutions must be determined by end user.
Research Category
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition

Linkage

Replaces: MAB3509

Physical form

Format: Purified
Purified immunoglobulin, by Protein G. Liquid in 10 mM PBS, pH 7,4 containing 0.2% BSA and 0.09% sodium azide.

Storage and Stability

Maintain at 2-8°C for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wanyuan Chen et al.
Oncology reports, 41(1), 67-76 (2018-10-27)
Pancreatic cancer (PC) is a lethal solid malignancy with resistance to traditional chemotherapy. Recently, considerable studies have demonstrated the ubiquitous antitumor properties of gene therapy mediated by the oncolytic vaccinia virus. The second mitochondrial‑derived activator of caspase (Smac) has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service