Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB411-C

Sigma-Aldrich

Anti-Interferon-α2 Antibody, clone ST254

clone ST254, from mouse

Synonym(s):

Interferon alpha-2, IFN-alpha-2, Interferon alpha-A, LeIF A, Interferon-α2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$464.00

$464.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$464.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$464.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ST254, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
affinity binding assay: suitable
neutralization: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... IFNA2(3440)

General description

Interferon alpha-2 (UniProt P01563; also known as IFN-alpha-2, Interferon alpha-A, LeIF A) is encoded by the IFNA2 (also known as IFNA2A, IFNA2B, IFNA2C) gene (Gene ID 3440) in human. Interferons (IFNs) are cytokines whose prouction and release by host cells are induced in the presence of pathogens (e.g. viruses, bacteria, parasites) and tumor cells. Originally named for their ability to interfere with viral replication, IFNs mediate the activation of immune cells, such as natural killer cells and macrophages, and increase host defense by up-regulating antigen presentation by boosting the expression of major histocompatibility complex (MHC) antigens. More than twenty distinct IFN genes and proteins have been identified and classified into three main types. Type I IFNs (IFN-α, IFN-β, IFN-ε, IFN-κ, and IFN-ω) exert their function via binding IFN-α/β receptor (encoded by the IFNAR1 and IFNAR2 gene, respectively). IFN-γ is the only known type II IFN, it is released by type 1 T helper cells and binds IFN-γ receptor (encoded by the IFNGR1 and IFNGR2 gene, respectively). Type III IFNs signal through a receptor complex consisting of IL10R2 (CRF2-4) and IFNLR1 (CRF2-12).
This Anti-Interferon-α2 Antibody, clone ST254 (Ascites Free) is validated for use in Western Blotting, Affinity Binding Assays, ELISA and Neutralizing.

Specificity

Clone ST254 neutralized human IFN-αA (IFN-α2) and IFN-αN (produced by Nalmalwa human Burkitt′s lymphoma cells), but not IFN-α1, antiviral activity (Shearer, M., et al. (1984). J. Immunol. 133(6):3096-3101).

Immunogen

Human alpha-interferon.

Application

Neutralizing Analysis: A representative lot enhanced viral replication by neutralizing host-secreted IFN in human prostate cancer PC3 cultures infected with P/V-CPI(−) and P/V-CPI(−)-G3A at a low MOI (Gainey, M.D., et al. (2000). J. Virol. 82(19):9369-9380).
Neutralizing Analysis: A representative lot neutralized human IFN-αA (IFN-α2) and IFN-αN (produced by Nalmalwa human Burkitt′s lymphoma cells), but not IFN-α1, antiviral activity against Semliki Forest Virus in human WISH amniotic epithelial cell cultures (Shearer, M., et al. (1984). J. Immunol. 133(6):3096-3101).
Affinity Binding Assay: A representative lot captured human IFN-α2, but not human IFN-α1 (Shearer, M., et al. (1984). J. Immunol. 133(6):3096-3101).
ELISA Analysis: A representative lot was used as the detection antibody for the detection of human IFN-αN (produced by Nalmalwa human Burkitt′s lymphoma cells) by sandwich ELISA (Shearer, M., et al. (1984). J. Immunol. 133(6):3096-3101).
Western Blotting Analysis: A representative lot detected IFN-αA (IFN-α2) and IFN-αN (produced by Nalmalwa human Burkitt′s lymphoma cells) under reducing condition by Western blotting (Shearer, M., et al. (1984). J. Immunol. 133(6):3096-3101).
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms
This Anti-Interferon-α2 Antibody, clone ST254 is validated for use in Western Blotting, Affinity Binding Assays, ELISA and Neutralizing

Quality

Evaluated by Polyacrylamide Gel Electrophoresis.

Purity of this monoclonal antibody is determined to be at least >85% by SDS-PAGE.

Target description

21 kDa calculated.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service