Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB3839

Sigma-Aldrich

Anti-Tissue Transglutaminase Antibody, FN binding domain, clone 4G3

clone 4G3, Chemicon®, from mouse

Synonym(s):

tTG, Protein-glutamine gamma-Glutamyltransferase, Transglutaminase 2, TGase C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
clone:
4G3, monoclonal
application:
ICC
species reactivity:
human
technique(s):
immunocytochemistry: suitable
citations:
6

biological source

mouse

antibody form

purified immunoglobulin

clone

4G3, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TGM2(7052)

General description

Tissue transglutaminase (tTG) is a 75 kDa monomeric globular protein expressed in the majority of cells and tissues. tTG localizes mainly in the cytoplasm but some tGT is present on the cell surface and is association with the extracellular matrix. In addition to its guanosine triphosphatase (GTPase) activity, tTG catalyzes the posttranslational modification of proteins by transamidation of available glutamine residues (Monsonego, 1997). Extracellular tTG is able to bind to and cross-link several ECM proteins and may function to stabilize the ECM and basement membranes. The protein is translated as a fully active enzyme and there is no evidence for proteolytic activation.

High constitutive expression and accumulation of active tTG is observed in (among others) endothelial cells, platelets, vascular smooth muscle cells and epithelial cells of the lens. tTG is expressed at very low levels in skeletal muscle cells and neurons and may be difficult to detect in these cell/tissue types.

Specificity

Tissue Transglutaminase. Will recognize all three known splice variants of tTG.

Immunogen

Epitope: FN binding domain
Fibronectin binding domain of tissue transglutaminase (amino acids 1-165 of tTG).

Application

Detect Tissue Transglutaminase using this Anti-Tissue Transglutaminase Antibody, FN binding domain, clone 4G3 validated for use in IC.
Immunohistochemistry: 20 μg/mL

Adhesion Blocking: 10-20 μg/mL (per 2x10e5 cells). 4G3 interferes with adhesion of TPA stimulated THP-1 cells and M-CSF stimulated peripheral blood monocytes to fibronectin.

Optimal working dilutions must be determined by the end user.



PROTOCOL:

IMMUNOCYTOCHEMISTRY: THP-1 cells were induced to differentiate into macrophages by treatment with 150 ng/ml of TPA and plated on fibronectin coated glass coverslips for 72 hours.Live non-permeablized cells were incubated with 20 μg/mL of MAB3839 for 45 minutes, washed three times with PBS and fixed for 10 minutes with 3.7% paraformaldehyde. The cells were then washed with PBS and incubated 45 minutes with a fluoresceine labeled affinity-purified goat-anti-mouse secondary antibody. After extensive washing with PBS, cells were mounted in Fluorsave medium.

Observed Staining:Cell surface tTG was colocalized with b1- and b3-Integrins in podosome-like adhesive structures of macrophages adherent on fibronectin.
Research Category
Apoptosis & Cancer

Metabolism
Research Sub Category
Apoptosis - Additional

Enzymes & Biochemistry

Physical form

Format: Purified
Protein A purified immunoglobulin in 0.02M Phosphate Buffer, pH 7.6, 0.25M NaCl containing 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Antonella Nadalutti et al.
PloS one, 8(10), e77277-e77277 (2013-10-17)
To investigate the role of thioredoxin (TRX), a novel regulator of extracellular transglutaminase 2 (TG2), in celiac patients IgA (CD IgA) mediated TG2 enzymatic activation. TG2 enzymatic activity was evaluated in endothelial cells (HUVECs) under different experimental conditions by ELISA
Ellen A Rorke et al.
Molecular carcinogenesis, 61(1), 19-32 (2021-10-06)
Type 2 transglutaminase (TG2) functions as an important cancer cell survival protein in a range of cancers including epidermal squamous cell carcinoma. TG2 exists in open and closed conformations each of which has a distinct and mutually exclusive activity. The
C Kerr et al.
Oncogene, 36(21), 2981-2990 (2016-12-13)
Type 2 transglutaminase (TG2) is an important cancer stem cell survival protein that exists in open and closed conformations. The major intracellular form is the closed conformation that functions as a GTP-binding GTPase and is required for cancer stem cell
Gautam Adhikary et al.
Oncotarget, 9(77), 34495-34505 (2018-10-24)
Mesothelioma is a rare cancer of the mesothelial cell layer of the pleura, peritoneum, pericardium and tunica vaginalis. It is typically caused by asbestos, notoriously resistant to chemotherapy and generally considered incurable with a poor life expectancy. Transglutaminase 2 (TG2)
Warren Naselsky et al.
Molecular carcinogenesis, 61(6), 537-548 (2022-03-24)
Transglutaminase 2 (TG2) is an important mesothelioma cancer cell survival protein. However, the mechanism whereby TG2 maintains mesothelioma cell survival is not well understood. We present studies showing that TG2 drives hepatocyte growth factor (HGF)-dependent MET receptor signaling to maintain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service