Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB3468

Sigma-Aldrich

Anti-β Galactosidase Antibody, clone DC1-4C7

ascites fluid, clone DC1-4C7, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

DC1-4C7, monoclonal

species reactivity

E. coli, vertebrates

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GLB1(2720)

Specificity

Beta galactosidase.

Immunogen

Beta galactosidase from E. coli.

Application

Anti-β Galactosidase Antibody, clone DC1-4C7 is an antibody against β Galactosidase for use in ELISA & WB.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags
Western blot

ELISA

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: MAB1802

Physical form

Liquid. Contains no preservative.

Storage and Stability

Maintain at -20°C in undiluted aliquots up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional and topological analysis of the Burkholderia cenocepacia priming glucosyltransferase BceB, involved in the biosynthesis of the cepacian exopolysaccharide.
Videira, PA; Garcia, AP; Sa-Correia, I
Journal of Bacteriology null
Eran Perlson et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(31), 9903-9917 (2009-08-07)
Retrograde axonal transport of cellular signals driven by dynein is vital for neuronal survival. Mouse models with defects in the retrograde transport machinery, including the Loa mouse (point mutation in dynein) and the Tg(dynamitin) mouse (overexpression of dynamitin), exhibit mild

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service