Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB335

Sigma-Aldrich

Anti-Synaptobrevin Antibody, clone SP11

clone SP11, Chemicon®, from mouse

Synonym(s):

VAMP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

SP11, monoclonal

species reactivity

human, feline, pig

should not react with

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... VAMP1(6843)

Specificity

Synaptobrevin [VAMP], 16kDa protein. Not reactive with bacterially expressed fusion protein.

Application

Immunohistochemistry at 1:100-1:200

Formalin fixed (fixed <4 weeks), paraffin embedded

Frozen

4% Paraformaldehyde

Western blotting at 1:100-1:1,000.

ELISA at 1:500

Optimal working dilutions must be determined by the end user.
This Anti-Synaptobrevin Antibody, clone SP11 is validated for use in ELISA, WB, IH(P) for the detection of Synaptobrevin.

Physical form

Format: Purified
Liquid in 0.02 MPB and 0.25M NaCl, PH 7.6 with 0.1% sodium azide.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service