Skip to Content
MilliporeSigma
All Photos(2)

Documents

MAB318-AF555

Sigma-Aldrich

Anti-Tyrosine Hydroxylase Antibody, clone LNC1, Alexa Fluor 555 Conjugate

clone LNC1, from mouse, ALEXA FLUOR 555

Synonym(s):

Tyrosine 3-monooxygenase, Dystonia 14, Tyrosine 3-hydroxylase, TH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 555

antibody form

purified antibody

antibody product type

primary antibodies

clone

LNC1, monoclonal

species reactivity

chicken, zebrafish, mouse, rat, frog, vole, monkey, human

technique(s)

immunocytochemistry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TH(7054)
rat ... Th(25085)

General description

Tyrosine 3-monooxygenase (EC 1.14.16.2; UniProt P07101; also known as Dystonia 14, TH, Tyrosine 3-hydroxylase) is encoded by the TH (also known as DYT14, DYT5b, TYH) gene (Gene ID 7054) in human. Tyrosine hydroxylase plays an important role in the physiology of adrenergic neurons. It is the first and rate-limiting enzyme involved in the biosynthesis of dopamine and norepinephrine from tyrosine. TH is, therefore, a useful marker for dopaminergic and noradrenergic neurons. TH enzymatic activity requires ferrous ions as cofactor and is believed to be regulated by phosphorylation. Six isoforms of human TH exist as a result of alternative splicing (UniProt P07101-1 through P07101-6).

Specificity

Recognizes an epitope on the outside of the regulatory N-terminus. The unconjugated antibody (Cat. No. MAB318) recognizes a protein of approximately 59-61 kDa by Western blotting. The unconjugated antibody does not react with the following by Western blotting: dopamine-beta-hydroxylase, phenylalanine hydroxylase, trytophan hydroxylase, dehydropteridine reductase, sepiapterin reductase or phenethanolamine-N-methyl transferase (PNMT).

Immunogen

Epitope: On the outside of the regulatory N-terminus of tyrosine hydroxylase.
Tyrosine Hydroxylase purified from PC12 cells.

Application

Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers
The unconjugated antibody (Cat. No. MAB318) is shown to be suitable also for Western blotting, immunohistochemistry, and immunoprecipitation applications.

Quality

Evaluated by Immunocytochemistry in rat cortical neurons.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Tyrosine Hydroxylase in rat cortical neurons.

Target description

~59-63 kDa observed.

Physical form

Protein A purified
Purified mouse monoclonal IgG1κ antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service