Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAB3150-M

Sigma-Aldrich

Anti-Cardiac Troponin I, a.a. 41-49, clone 284 (19C7) (1mg) KC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

Quality Level

clone

284
monoclonal

species reactivity

( Bovine Canine Goat Human Mouse Pig Rat Rabbit Fish Feline ), bovine, mouse

technique(s)

ELISA: suitable
western blot: suitable

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service