Skip to Content
MilliporeSigma
All Photos(2)

Documents

MAB2623

Sigma-Aldrich

Anti-EMMPRIN [CD147] Antibody, clone 1S9-2A

clone 1S9-2A, from mouse

Synonym(s):

Basigin, CD147 antigen, Collagenase stimulatory factor, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor, TCSF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1S9-2A, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BSG(682)

General description

Matrix metalloproteinases (MMPs) or gelatinases play an important role in the early invasion of malignant tumors. These enzymes may be released directly from tumor cells, or may be derived from fibroblasts that have been stimulated with the extracellular MMP-inducer EMMPRIN. Studies have demonstrated that EMMPRIN stimulates human fibroblast production of interstitial collagenase, stromelysin-1, and gelatinase A (MMP-2), which is essential for tissue degradation and remodeling during tumor invasion and wound healing.

Specificity

The antibody recognizes EMMPRIN.

Immunogen

Epitope: Unknown
Human synovial fibroblasts.

Application

Imunohistochemistry Analysis: A 1:100 dilution from a previous lot detected EMMPRIN in human adenocarcinoma tissue.
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs
This Anti-EMMPRIN [CD147] Antibody, clone 1S9-2A is validated for use in WB, IH for the detection of EMMPRIN [CD147].

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.1 µg/ml of this antibody detected EMMPRIN on 10 µg of HeLa cell lysate.

Target description

~ 35-58 kDa The extent of CD147 glycosylation is tissue-specific and is responsible for a variation in the apparent molecular mass from 35 kDa to 58 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Larissa Sweeny et al.
Journal of cutaneous pathology, 39(6), 603-609 (2012-05-12)
CD147 is upregulated in multiple cancer types, but its expression in advanced cutaneous squamous cell carcinoma (SCC) is unknown. Our purpose was to evaluate the expression patterns of CD147 and related monocarboxylate transporters (MCT1, MCT4) to determine their correlation with
Barrett L Updegraff et al.
Cell reports, 25(8), 2223-2233 (2018-11-22)
Pathways underlying metabolic reprogramming in cancer remain incompletely understood. We identify the transmembrane serine protease TMPRSS11B as a gene that promotes transformation of immortalized human bronchial epithelial cells (HBECs). TMPRSS11B is upregulated in human lung squamous cell carcinomas (LSCCs), and high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service