Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB2291

Sigma-Aldrich

Anti-Integrin α3 Antibody, cytoplasmic domain, clone 54B3

clone 54B3, Chemicon®, from mouse

Synonym(s):

CD49c

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

54B3, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA3(3675)

General description

Integrins are a family of heterodimeric membrane glycoproteins consisting of non-covalently associated alpha and beta subunits. More than 18 alpha and 8 beta subunits with numerous splice variant isoforms have been identified in mammals. In general, integrins function as receptors for extracellular matrix proteins. Certain integrins can also bind to soluble ligands or to counter-receptors on adjacent cells, such as the intracellular adhesion molecules (ICAMs), resulting in aggregation of cells. Signals transduced by integrins play a role in many biological processes, including cell growth, differentiation, migration and apoptosis. For integrin subunits alpha 3 and alpha 6, two cytoplasmic variants, A and B, have been identified.
MAB2291 is a mouse monoclonal generated to a synthetic peptide in the cytoplasmic domain of integrin alpha 3B.

Specificity

Anti-Integrin alpha 3B recognizes the cytoplasmic domain of integrin alpha 3B. Integrin alpha 3B is present in the microvascular structures in brain and heart.

SPECIES REACTIVITIES:

Although untested, a broad species reactivity is expected due to the conserved nature of the epitope.

Immunogen

Epitope: cytoplasmic domain
Synthetic peptide: CTRYYQIMPKYHAVRIREEERYPPPGSTLPTKK

Application

Research Category
Cell Structure
Research Sub Category
Integrins
This Anti-Integrin α3 Antibody, cytoplasmic domain, clone 54B3 is validated for use in WB, IC, IH for the detection of Integrin α3.
Western Blotting

Immunocytochemistry

Immunohistochemistry

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Purified. Liquid in PBS containing 0.1% sodium azide.

Storage and Stability

Store at 2-8°C, or in small aliquots at -20°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service