Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1992

Sigma-Aldrich

Anti-Integrin α3β1 Antibody, clone M-KID2

clone M-KID2, Chemicon®, from mouse

Synonym(s):

VLA-3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$564.00

$564.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1389

Request a Bulk Order

Select a Size

Change View
100 μG
$564.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$564.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1389

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

M-KID2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
radioimmunoassay: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA3(3675)

General description

Integrins are a family of dimeric proteins that mediate cell-cell and extracellular matrix adhesion. Each VLA subtype (VLA-1, VLA-2, VL-3, VLA-4, VLA-5) is composed of a distinct alpha subunit noncovalently associated with a common beta subunit, as an alpha-beta dimer. Integrin VLA-3 (very late antigen, alpha-3/beta-1) is a cell surface receptor that binds to fibronectin, laminin, collagen type I and VI. VLA-3 is typically restricted in normal tissue but is widely expressed in many cultured tumor cells and correlates with the degree of invasiveness of malignant melanoma cells.

Specificity

Recognizes an epitope related to the VLA-3 complex (alpha-3/beta-1 integrin). Specificity was determined by immunoprecipitation from a detergent extract of 35S-methionine labeled KJ29 cells.

Immunogen

Human renal carcinoma cell line (KJ29).

Application



Immunohistochemistry (works on Acetone-fixed paraffin-embedded tissues)

ELISA/RIA

Immunoprecipitation. When immunoprecipitated samples are run on SDS-PAGE under a) non-reducing conditions, two bands will be detected with an apparent molecular weight of 110 and 150kDa; and b) reducing (2% B-ME) conditions, a single molecular species of 130kDa is detected. Under reducing conditions both alpha3 and beta1 subunits run with the same apparent molecular weight.

Optimal working dilutions must be determined by end user.
Anti-Integrin α3β1 Antibody, clone M-KID2 detects level of Integrin α3β1 & has been published & validated for use in ELISA, IP, RIA, IC, IH(P).

Physical form

Format: Purified
Immunoglobulin purified by Protein A. Purified immunoglobulin in PBS and 0.05% sodium azide as preservative.

Storage and Stability

Maintain at 2°-8°C in undiluted aliquots for up to 12 months from date of receipt.

Analysis Note

Control
Cell line: KJ29 renal carcinoma adherent cells.
Tissue: Kidney; antibody reacts with basal layer podocytes and basement membrane of distal tubules.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehrnoosh Saghizadeh et al.
Experimental eye research, 129, 66-73 (2014-12-03)
Overexpression of c-met and suppression of matrix metalloproteinase-10 (MMP-10) and cathepsin F genes was previously shown to normalize wound healing, epithelial and stem cell marker patterns in organ-cultured human diabetic corneas. We now examined if gene therapy of limbal cells
Y Nakamura et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 49(4), 439-444 (2001-03-22)
The tetra-membrane-spanning protein CD9 forms a complex with a membrane-anchored heparin binding epidermal growth factor-like growth factor (HB-EGF) and integrin alpha3beta1 in some human and monkey cell lines. We show here the immunohistochemical distribution of CD9, HB-EGF, and integrin alpha3beta1
Sara Brilha et al.
Frontiers in immunology, 9, 1348-1348 (2018-07-11)
Pulmonary tuberculosis (TB) is caused by inhalation of Mycobacterium tuberculosis, which damages the bronchial epithelial barrier to establish local infection. Matrix metalloproteinase-1 plays a crucial role in the immunopathology of TB, causing breakdown of type I collagen and cavitation, but
Enhanced wound healing, kinase and stem cell marker expression in diabetic organ-cultured human corneas upon MMP-10 and cathepsin F gene silencing.
Saghizadeh, M; Epifantseva, I; Hemmati, DM; Ghiam, CA; Brunken, WJ; Ljubimov, AV
Investigative Ophthalmology & Visual Science null
T Takeuchi et al.
The Journal of clinical investigation, 92(6), 3008-3016 (1993-12-01)
Upregulation of integrin adhesive receptors has been implicated in various pathological conditions. We examined expression and function of integrin adhesive receptors on peripheral blood lymphocytes from patients with systemic lupus erythematosus (SLE), particularly those with the complication of vasculitis, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service