Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1920-C

Sigma-Aldrich

Anti-Laminin γ1 Antibody, clone 2E8 (Ascites Free)

clone 2E8, from mouse

Synonym(s):

Laminin subunit gamma-1, Laminin B2 chain, Laminin-1/-2/-3/-4/-6/-7/-8/-9/-10/-11 subunit gamma, S-laminin subunit gamma, S-LAM gamma

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$599.00

$599.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$599.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$599.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E8, monoclonal

species reactivity

mouse, rat, feline, human

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... LAMC1(3915)

General description

Laminin subunit gamma-1 (UniProt P11047; also known as Laminin B2 chain, Laminin-1/-2/-3/-4/-6/-7/-8/-9/-10/-11 subunit gamma, S-laminin subunit gamma, S-LAM gamma) is encoded by the LAMC1 (also known as LAMB2) gene (Gene ID 3915) in human. Laminins are basement membrane, extracellular matrix glycoproteins composed of alpha, beta, and gamma heterotrimeric chains. Different combinations of five alpha chains, three beta chains, and three gamma chains form the 15 known laminin heterotrimers, nine of which contain the gamma-1 chain: Laminin-1/EHS (α1β1γ1), Laminin-2/Merosin (α2β1γ1), Laminin-3/S-laminin (α1β2γ1), Laminin-4/S-merosin (α2β2γ1), Laminin-6/K-laminin (α3β1γ1), Laminin-7 (α3β2γ1), Laminin-8 (α4β1γ1), Laminin-9 (α4β2γ1), Laminin-10 (α5β1γ1), Laminin-11 (α5β2γ1). The gamma chain is initially produced with a signal peptide (a.a. 1-33), the removal of which yields the 1576-amino acid mature protein (a.a. 34-1609) with 11 EGF-like domains and a Laminin IV type A domain. Laminins are associated with type IV collagen networks via entactin, fibronectin, and perlecan. They also bind to cell membranes through integrin receptors and other plasma membrane molecules, such as the dystroglycan glycoprotein complex and Lutheran blood group glycoprotein. Laminins play important roles in cell attachment and differentiation, cell shape and movement, maintenance of tissue phenotype and survival.

Specificity

Detects the laminin γ1 component of Laminin-1/EHS (α1β1γ1), Laminin-2/Merosin (α2β1γ1), Laminin-3/S-laminin (α1β2γ1), Laminin-4/S-merosin (α2β2γ1), Laminin-6/K-laminin (α3β1γ1), Laminin-7 (α3β2γ1), Laminin-8 (α4β1γ1), Laminin-9 (α4β2γ1), Laminin-10 (α5β1γ1), Laminin-11 (α5β2γ1).

Immunogen

Epitope: Center of the laminin cross.
Human placenta laminin pepsin digest.

Application

Anti-Laminin γ1 Antibody, clone 2E8 (Ascites Free) is an antibody against Laminin γ1 for use in Western Blotting, Immunofluorescence, Immunocytochemistry, ELISA.
Immunofluorescence Analysis: A representative lot detected laminin γ1 immunoreactivity in the extracellular matrix fractones by fluorescent immunohistochemistry using human and mouse brain cryosections (Kerever, A., et al. (2007). Stem Cells. 25(9):2146-2157).
Immunofluorescence Analysis: A representative lot immunostained the seminiferous cord basement membrane and vasculature in frozen rat testis cryosections by fluorescent immunohistochemistry (Johnson, K.J., et al. (2007). Biol. Reprod. 77(6):978-989).
Immunofluorescence Analysis: A representative lot detected Laminin γ1 immunoreactivity in unfixed frozen cat muscle sections by fluorescent immunohistochemistry (O′Brien, D.P., et al. (2001). J.Neurol. Sci. 189(1-2):37-43).
Immunofluorescence Analysis: A representative lot immunostained the trophoblast and the blood vessel basement membranes in frozen human placenta tissue sections by fluorescent immunohistochemistry (Engvall, E., et al. (1986). J. Cell Biol. 103(6 Pt 1):2457-2465).
Immunocytochemistry Analysis: A representative lot detected the laminin γ1 immunoreactivity at the substratum-attached surface of formaldehyde-fixed, Triton X-100-permeablized rat lung aveolar epithelial cells (AECs) by fluorescence immunocytochemistry (Jones, J.C., et al. (2005). J. Cell Sci. 118(Pt 12):2557-2566).
Western Blotting Analysis: Representative lots detected the ~200 kDa target band using rat lung aveolar epithelial cell (AEC) lysates, human amnion extracts, and rat yolk sac tumors (Jones, J.C., et al. (2005). J. Cell Sci. 118(Pt 12):2557-2566; Engvall, E., et al. (1986). J. Cell Biol. 103(6 Pt 1):2457-2465).
ELISA Analysis: A representative lot detected the binding of human laminin-10 (α5β1γ1) to immobilized α-dystroglycan and heparin by ELISA (Ido, H., et al. (2004). J. Biol. Chem. 279(12):10946-10954).
ELISA Analysis: A representative lot detected laminin γ1 subunit in human and rat, but not mouse, laminin preparations by ELISA (Engvall, E., et al. (1986). J. Cell Biol. 103(6 Pt 1):2457-2465).
Immunohistochemistry Analysis: A representative lot detected Laminin γ1 immunoreactivity in the perineurium of the perineurial cell basement membrane (PCBM) layers in paraffin-embedded human sural nerve sections (Hill, R.E., and Williams, R.E. (2002). J. Anat. 201(2):185-192).
Immunoprecipitation Analysis: A representative lot immunoprecipitated laminin from the culture medium of human choriocarcinoma (JAr) cells (Engvall, E., et al. (1986). J. Cell Biol. 103(6 Pt 1):2457-2465).
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Western Blotting in human heart tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Laminin γ1 in 10 µg of human heart tissue lysate.

Target description

~200 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service