Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1687

Sigma-Aldrich

Anti-Vimentin Antibody, clone 43BE8

ascites fluid, clone 43BE8, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

43BE8, monoclonal

species reactivity

human, monkey

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)

isotype

IgM

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... VIM(7431)

Specificity

Vimentin (58kD). Reacts in immunocytochemical assays with all mesenchymal cells (fibroblasts, Scwann cells, endothelial cells, smooth muscle cells, etc.).

Application

Immunohistochemistry: 1:500-1:2,000 on paraffin embedded sections (ABC peroxidase) fixed with alcohol (e.g. Carnoy) with a 2" pronase digestion. Aldehyde fixation is not recommended.

Immunocytochemistry: 1:100-1:200 cells in vitro (IFA) fixed in ice cold absolute methanol or acetone.

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton
This Anti-Vimentin Antibody, clone 43BE8 is validated for use in IC, IH(P) for the detection of Vimentin.

Physical form

Liquid

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amir Bajouri et al.
Cell journal, 22(1), 75-84 (2019-10-14)
Recently, the promising potential of fibroblast transplantation has become a novel modality for skin rejuvenation. We investigated the long-term safety and efficacy of autologous fibroblast transplantation for participants with mild to severe facial contour deformities. In this open-label, single-arm phase
Skin Rejuvenation with Cultured Melanocyte and Fibroblasts in a Medical Tourism Patient.
Atefeh Shahbazi et al.
World journal of plastic surgery, 11(3), 116-121 (2023-01-26)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service