Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1620-AF647

Sigma-Aldrich

Anti-Cytokeratin 5,6 Antibody, clone D5/16B4, Alexa Fluor 647

clone D5/16B4, from mouse, ALEXA FLUOR 647

Synonym(s):

Keratin, type II cytoskeletal 5, 58 kDa cytokeratin, Cytokeratin-5, CK-5, Keratin-5, K5, Type-II keratin Kb5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

conjugate

ALEXA FLUOR 647

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

D5/16B4, monoclonal

species reactivity

mouse, human, rat

technique(s)

immunocytochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KRT5(3852)

General description

Keratin, type II cytoskeletal 5 (UniProt P13647; also known as 58 kDa cytokeratin, CK-5, Cytokeratin-5, K5, Keratin-5, Type-II keratin Kb5) is encoded by the KRT5 (also known as DDD1, DM-EBS, EBSMCE, K-EBS, MP-EBS, WC-EBS) gene (Gene ID 3852) in human. Cytokeratin-5 (Keratin-5 or K5) belongs to a family of highly conserved epithelial-specific intermediate filament (IF) proteins. Each epithelium is characterized by its unique keratin expression pattern. In interfollicular epidermis, four major keratins are synthesized and co-polymerize in pairs, K5/K14 and K1/KI0, to form filaments. The expression of these two pairs is strictly regulated during normal keratinocyte differentiation. The K5/K14 filament is expressed in proliferating basal cells, while the K1/K10 filament is expressed during the transition to terminal differentiation and at the cessation of cell division. Similar to other IF members, Keratin-5 contains a central rod region composed of alpha helix segments (a.a. 168-203, 223-315, 339-477) that allow two different IF proteins, one type I (acidic) and one type II (basic) keratin, to intertwine into a coiled-coil formation via hydrophobic interaction. Kearain-5 is a type II keratin that pairs with the type I keratin-14 to form IF.

Specificity

Clone D5/16B4 detected cytokeratin 5 by Western blotting using cytoskeletal preparations from human epidermis and non-cytokeratinizing epithelium (Abstract: European Symposium of the Biology of the Cytoskeleton, Helsinki, 18-21, 6, 89, 1989), as well as by staining tissue sections. Clone D5/16B4 also shows reactivity toward cytokeratin 6 and weak reactivity with cytokeratin 4 by Western blotting. No cross-reactivity to cytokeratin 1, 7, 8, 10, 13, 14, 18 or 19. Clone D5/16B4 immunostains basal cells and a part of the stratum spinosum in the normal pavement epithelium. Clone D5/16B4 is suitable for staining formalin-fixed tissue sections.

Immunogen

Purified cytokeratin 5 (Mischke, D., and Wild, G.A. (1987). J. Invest. Dermatol. 88(2):191-197; Wild, G.A., and Mischke, D. (1986). Exp. Cell Res. 162(1):114-126).

Application

Anti-Cytokeratin 5,6 Antibody, clone D5/16B4, Alexa Fluor 647 is an antibody against Cytokeratin 5,6 for use in Immunocytochemistry.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton
The unconjugated antibody (Cat. No. MAB1620) is shown to be suitable also for immunofluorescence, immunohistochemistry, and Western blotting applications.

Quality

Evaluated by Immunocytochemistry in A431 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Cytokeratin 5/6 in A431 cells.

Target description

Cytokeratin 4 (59 kDa), Cytokeratin 5 (58 kDa) and Cytokeratin 6 (56 kDa).

Physical form

Protein A purified
Purified mouse monoclonal IgG1 antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service