Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAB1570

Sigma-Aldrich

Anti-NMDAR1 Antibody, CT, C2 cassette, clone 2C6.2

clone 2C6.2, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C6.2, monoclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2b

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Specificity

NMDAR1, C-terminal, C2 cassette

Immunogen

Epitope: C-terminus, C2 cassette
Synthetic peptide (LQNQKDTVLPRRAIEREEGQLQLCSRHRES) corresponding to the C-terminus of rat NMDA receptor subunit.

Application

Anti-NMDAR1 Antibody, C-terminus, C2 cassette, clone 2C6.2 is an antibody against NMDAR1 for use in IH & WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot: 1:100-1:500

Immunohistochemistry: 1:100-1:500 using paraformaldehyde or paraformaldehyde/glutaraldehyde fixed tissue with light and electron microscopy.

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02M Phosphate buffer, 0.25 M NaCl with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tom J Phillips et al.
Scientific reports, 7(1), 9079-9079 (2017-08-24)
Some neuropsychiatric disease, including schizophrenia, may originate during prenatal development, following periods of gestational hypoxia and placental oxidative stress. Here we investigated if gestational hypoxia promotes damaging secretions from the placenta that affect fetal development and whether a mitochondria-targeted antioxidant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service