Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1288-M

Sigma-Aldrich

Anti-HLA-B7 Antibody, clone BB7.1, PE Conjugate

clone BB7.1, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BB7.1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... HLA-B(3106)

General description

BIOCHEMISTRY: p I = 6.3

Specificity

Binds to the HLA-B7 antigen. It does not bind to HLA-B27 or other related HLA antigens.

Cell type Reactivity
Homozygous HLA-B7 ++
Heterozygous HLA-B7 +
Heterozygous HLA-B27 -
Non-HLA-B7, B27 -

Immunogen

Papain solubilized HLA-B7 antigen1

Application

Flow cytometry - Use 10 μL direct from the vial per 100 μL of whole blood, or 1 x 10E6 peripheral blood mononuclear cells (PBMC) in 100 μL buffer.

Optimal working concentrations must be determined by the end user.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-HLA-B7 Antibody, clone BB7.1, PE Conjugate is validated for use in FC for the detection of HLA-B7.

Physical form

Protein A purified
Purified mouse immunoglobulin in phosphate buffered saline, pH 7.4, containing 0.2% bovine Serum albumin and 0.1% sodium azide.

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Widely expressed

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monoclonal antibodies for analysis of the HLA system.
F M Brodsky et al.
Immunological reviews, 47, 3-61 (1979-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service