Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1288

Sigma-Aldrich

Anti-HLA-B7 Antibody, clone BB7.1, PE Conjugate

clone BB7.1, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BB7.1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HLA-B(3106)

Related Categories

General description

BIOCHEMISTRY: p I = 6.3
HLA-B belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a membrane-anchored heavy chain and a light chain (beta-2 microglobulin). HLA class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen and are expressed in nearly all cells. HLA-B7 is an HLA-B serotype. B7, previously HL-A7, is a serotype that identifies the more common HLA-B*07 gene products and was one of the first ′HL-A′ antigens recognized. B7 is a risk factor for cervical cancer, sarcoidosis, and early-onset spondylarthropathies.
The previously assigned protein identifier P03989 has been merged into P01889. Full details can be found on the UniProt database.

Specificity

Binds to the HLA-B7 antigen. It does not bind to HLA-B27 or other related HLA antigens.

Cell type Reactivity
Homozygous HLA-B7 ++
Heterozygous HLA-B7 +
Heterozygous HLA-B27 -
Non-HLA-B7, B27 -

Immunogen

Papain solubilized HLA-B7 antigen1

Application

Flow cytometry - Use 10 μL direct from the vial per 100 μL of whole blood, or 1 x 10E6 peripheral blood mononuclear cells (PBMC) in 100 μL buffer.

Optimal working concentrations must be determined by the end user.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-HLA-B7 Antibody, clone BB7.1, PE Conjugate is validated for use in FC for the detection of HLA-B7.

Physical form

Protein A purified
Purified mouse immunoglobulin in phosphate buffered saline, pH 7.4, containing 0.2% bovine Serum albumin and 0.1% sodium azide.

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Widely expressed

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ingrid M Overes et al.
Cancer immunology, immunotherapy : CII, 58(3), 429-439 (2008-08-23)
CD8(+) T cells recognizing minor histocompatibility antigens (MiHA) on solid tumor cells may mediate effective graft-versus-tumor (GVT) reactivity after allogeneic stem cell transplantation (SCT). Previously, we identified LRH-1 as a hematopoietic-restricted MiHA encoded by the P2X5 gene. Here, we report
Stefanie Ameres et al.
Journal of immunology (Baltimore, Md. : 1950), 192(12), 5894-5905 (2014-05-09)
Immunoevasive proteins ("evasins") of human CMV (HCMV) modulate stability and localization of MHC class I (MHC I) molecules, and their supply of antigenic peptides. However, it is largely unknown to what extent these evasins interfere with recognition by virus-specific CD8

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service