Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1087-I

Sigma-Aldrich

Anti-Eosinophil Peroxidase Antibody, clone AHE-1

Synonym(s):

EPO, EPX

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$202.00
100 μG
$508.00

$202.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$202.00
100 μG
$508.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$202.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

AHE-1, monoclonal

mol wt

calculated mol wt 81.04 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... EPX(8288)

General description

Eosinophil peroxidase (UniProt: P11678; also known as EC:1.11.1.7, EPO, EPX) is encoded by the EPX (also known as EPER, EPO, EPP) gene (Gene ID: 8288) in human. Eosinophil peroxidase is an enzyme found in cytoplasmic granules of eosinophils. It mediates tyrosine nitration of secondary granule proteins in mature resting eosinophils. It is synthesized with a signal peptide (aa 1-17) and a propeptide (aa 18-139), which are subsequently cleaved off to produce the mature form that can be cleaved into a light chain (aa 140-250) and a heavy chain (aa 251-715). These chains can assemble to for a tetrameric structure. It is a heme-containing enzyme that physically associates with fibrous tissue and cancer tissue in various organs. It contains well-conserved profibrogenic capacity to stimulate the migration of fibroblastic cells and promote their ability to secrete collagenous proteins to generate a functional extracellular matrix (ECM) both in vitro and in vivo. It plays a role in regulating the recruitment of fibroblast and the biosynthesis of collagen ECM at sites of normal tissue repair and fibrosis. It is a key participant in generation of reactive oxidants and diffusible radical species by the phagocytes. It serves as a potent toxin towards invading parasites and the surrounding tissues. It also displays significant inhibitory activity towards Mycobacterium tuberculosis H37Rv by inducing bacterial fragmentation and lysis. (Ref.: DeNichilo, MO., et al. (2015). Am. J. Pathol. 185(5); 1372-1384; Mitra, SN., et al. (2000). Redox Rep. 5(4); 215-224).

Specificity

Clone AHE-1 is a mouse monoclonal antibody that detects Eosinophil Peroxidase.

Immunogen

Human Eosinophils from a patient with hypereosinophilic syndrome.

Application

Quality Control Applications

Evaluated by Immunofluorescence in frozen Human colon tissue sections.

Immunofluorescence Analysis: A 1:50 dilution of this antibody detected Eosinophil Peroxidase in frozen Human colon tissue sections.

Tested applications

Immunofluorescence Analysis: A representative lot detected Eosinophil Peroxidase in Immunofluorescence applications (DeNichilo, M.O., et al. (2015). Am J Pathol. 185(5); 1372-84; Song, Y., et al. (2013). ISRN Inflamm. 2013; 907821).

Immunohistochemistry Applications: A representative lot detected Eosinophil Peroxidase in Immunohistochemistry applications (Spang, C., et al. (2017). J Musculoskelet Neuronal Interact. 17(3); 226-236; Song, Y., et al. (2013). ISRN Inflamm. 2013; 907821; Song, Y., et al. (2013). BMC Musculoskelet Disord. 14:; 34; Song, Y., et al. (2012). PLoS One ;7(12); e52230).

Immunocytochemistry Analysis: A representative lot detected Eosinophil Peroxidase in Immunocytochemistry applications (DeNichilo, M.O., et al. (2015). Am J Pathol. 185(5); 1372-84).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service