Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

M8CMNFX00

Millipore

Multiscreen® 96 well Plate, solid bottom

flat bottom clear polystyrene plate, non-sterile, pkg of 100 plates, medium binding surface

Synonym(s):

Solid Bottom Microplate

Sign Into View Organizational & Contract Pricing

Select a Size

100 EACH
$1,440.00

$1,440.00


Please contact Customer Service for Availability


Select a Size

Change View
100 EACH
$1,440.00

About This Item

UNSPSC Code:
41116158
NACRES:
NB.15
material:
flat bottom clear polystyrene plate
size:
96 well
sterility:
non-sterile
binding type:
medium binding surface

$1,440.00


Please contact Customer Service for Availability

material

flat bottom clear polystyrene plate

Quality Level

sterility

non-sterile

product line

MultiScreen®

packaging

pkg of 100 plates

technique(s)

ELISA: suitable

plate size

96 wells

size

96 well

binding type

medium binding surface

General description

MultiScreen® 96-well ELISA microplates meet industry standard dimensions for use in common biochemical assay platforms, including automated liquid handling, microplate readers, and other microplate compatible devices. ELISA microplates are available with medium or high binding capabilities.

Physical form

Color Code:Clear

Other Notes

Automation Compatible:Yes

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service