Key Documents
LX2RCAL-K25
Millipore
Luminex 200 Calibration Kit, 25 uses
About This Item
UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
HB.01
Pricing and availability is not currently available.
product line
Milliplex®
technique(s)
multiplexing: suitable
General description
All xMAP Instruments require regular calibration and performance verification testing, to ensure the system is operating correctly and to maintain data accuracy. The Luminex 100/200 instruments, operating on xPONENT software, are calibrated using a calibration kit and a verification kit, each providing 25 uses.
Application
For use with Luminex 200 system operating on xPONENT software
Other Notes
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial
Legal Information
Luminex 200 is a trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany