Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

LUXMIXTB

Millipore

mPAGE® Lux Mixing Tube

greener alternative

Opaque tube for mixing MPAGE® Lux reagents

Synonym(s):

opaque mixing tube

Sign Into View Organizational & Contract Pricing

Select a Size

5 EACH
$12.70

$12.70


Estimated to ship onApril 22, 2025Details


Request a Bulk Order

Select a Size

Change View
5 EACH
$12.70

About This Item

UNSPSC Code:
41105305
NACRES:
NB.11

$12.70


Estimated to ship onApril 22, 2025Details


Request a Bulk Order

Product Name

mPAGE® Lux Mixing Tube, 5pk,

material

high-density polyethylene lid
polypropylene tube

description

for use with mPAGE® Lux Curing Station

packaging

pack of 5 packs

manufacturer/tradename

mPAGE®

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

SDS-PAGE: suitable
electrophoresis: suitable

L × W × H

22.86 cm × 15.24 cm × 3.6 cm

compatibility

for use with

  • mPAGE® Lux Bis-Tris Reagent Kit
  • mPAGE® Lux Curing Station

greener alternative category

General description

Use mPAGE® Lux Mixing Tube, 5pk when mixing mPAGE® Lux Resolving Solution and mPAGE® Lux Diluent to avoid light.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service