Skip to Content
MilliporeSigma
All Photos(3)

Documents

LI0685BC

Millipore

TRANSIA® PLATE Listeria spp. 10X

BioControl, ELISA test for detection of Listeria in food and environmental samples

Synonym(s):

Listeria detection kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NA.85

Agency

AFNOR

product line

TRANSIA®

shelf life

3‑24 mo.

packaging

pkg of 960 tests

manufacturer/tradename

BioControl Systems LI0685

technique(s)

ELISA: suitable

input

food(s)

application(s)

food and beverages
pathogen testing

storage temp.

2-8°C

suitability

Listeria spp.

General description

TRANSIA® PLATE Listeria spp. 10X is an ELISA test kit that incorporates proven antibody-antigen "sandwich" assay technology for the detection of Listeria species in both food and environmental samples. Highly specific antibodies coated to polystyrene microtiter plate wells capture and bind the target antigen, if present. A detection antibody linked to a conjugate enzyme is introduced to form an antibody/antigen/antibody sandwich. Finally, a substrate is added and converted by the conjugate enzyme to produce a color change indicating the presence of the target pathogen. The extensive AOAC and AFNOR validations and built-in positive and negative controls combine to provide users with the necessary confidence in the results. The efficient divisible 96-well format and available automation make it an ideal solution for high-volume testing.

Application

TRANSIA® PLATE Listeria is a kit for detection of the Listeria (L. monocytogenes, L. ivanovii, L. innocua, L. seeligeri, L. welshimeri, and L. grayi) in dairy products, meat products, seafood products, vegetables, and environmental samples. The assay reliably recovers and detects low levels of Listeria spp. in a variety of samples.

Features and Benefits

  • Detects all 6 species of Listeria providing a true indication of product and surface quality.
  • Positive and Negative controls supplied with the kit.

Components

  • 10 microplates with dividable microwell strips with 96 wells (12 microwell strips of 8 microwells).
  • Negative control: (bacterial antigens not belonging to the Listeria genus), ready-to-use, 2 x 20 mL.
  • Positive control: Listeria monocytogenes antigens, ready-to-use, 2 x 10mL.
  • Conjugate (contains peroxidase conjugated anti-Listeria antibodies), ready-to-use, 2 x 57 mL.
  • Wash buffer, 20 X concentration, 800 mL.
  • Substrate (urea H2O2), ready-to-use, 1 x 57 mL.
  • Chromogen (TMB), ready-to-use, 1 x 57 mL.
  • Stop solution (H2SO4), ready-to-use, 2 x 57 mL.

Principle

TRANSIA® PLATE Listeria is an ELISA test (Enzyme Linked Immuno Sorbent Assay) based on a two step sandwich-type reaction. The solid support of the reaction is a microtitration plate with dividable microwellstrips. Specific antibodies of flagellar antigens are grafted on the microtitration plate wells.

Testing Method

ELISA

Legal Information

TRANSIA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service