Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KZ-125ML-BK

Millipore

Anti-e (clone MS-16), FFMU

Anti-e from the clone MS-16, Human IgM, Intermediate for further manufacturing use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116102
NACRES:
NA.41

biological source

human

Quality Level

antibody form

unpurified

clone

MS-16, monoclonal

form

liquid

packaging

pkg of 125 mL

concentration

3-14 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

6.7-7.7

isotype

IgM

shipped in

ambient

storage temp.

2-8°C

General description

Standardized, concentrated cell culture supernatant, non-diafiltered, minimal pH buffering; For the formulation of Anti-e blood typing reagents or devices.

Broad specificity, reactions are influenced by the presence or absence of the C antigen which makes it similar to an Anti-Ce; Contains 0.1% (w/v) Sodium Azide; Shelf Life: 18 months from date of filtration.

Licensed by the US FDA under a shared manufacturing agreement

Application

Antibody intermediate for further manufacturing use
Production of an in vitro blood typing product or device

Legal Information

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Partially processed reagent for further manufacture of non-injectable products only. The absence of all viral antigens has not been determined. No US Standard of Potency. Do not use if turbid. Contains 0.1% (w/v) sodium azide.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service