Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IESN07852

Millipore

Immobilon® -E PVDF Blotting Sandwich

20 sandwiches, 7 cm x 8.4 cm, transfer membrane interleaved with blotting paper

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
$294.00

$294.00


Please contact Customer Service for Availability


Select a Size

Change View
20 EA
$294.00

About This Item

UNSPSC Code:
41105339
eCl@ss:
32031602
NACRES:
NB.23

$294.00


Please contact Customer Service for Availability

Product Name

Immobilon®-E Blotting Sandwich, 7 x 8.4 cm, Immobilon-E membrane interleaved with blotting paper, for standard tank transfer procedures. PVDF membrane wets out in water or transfer buffer.

material

PVDF membrane
paper membrane
plain filter
white filter

Quality Level

feature

easily wetted with water

manufacturer/tradename

Immobilon®

technique(s)

western blot: suitable

filter L × W

7.0 cm × 8.4 cm

pore size

0.45 μm pore size

compatibility

for use with India ink
for use with Ponceau-S red
for use with Sypro ruby
for use with Transillumination

detection method

chemiluminescent
colorimetric
radioactive

shipped in

ambient

General description

Filter Code: IEVH
Immobilon-E is the first PVDF membrane for Western blotting that can be wetted with with aqueous transfer buffer. This eliminates the alcohol pre-wet step required with other PVDF membranes.

Application

7 x 8.4 cm, Immobilon-E membrane interleaved with blotting paper, for standard tank transfer procedures. PVDF membrane wets out in water or transfer buffer.

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service