HX0770
Hydroxylamine Hydrochloride
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
Synonym(s):
Hydroxylamine Hydrochloride, Hydroxylammonium Chloride, Hydroxyl Ammonium Chloride
About This Item
vapor pressure
0.054 Pa ( 50 °C)
Assay
≥96%
form
crystals
pH
2.5-3.5 (20 °C, 50 g/L in H2O)
mp
154 °C
solubility
470 g/L
density
1.7 g/cm3 at 20.2 °C
shipped in
ambient
storage temp.
room temp
InChI
1S/ClH.H3NO/c;1-2/h1H;2H,1H2
InChI key
WTDHULULXKLSOZ-UHFFFAOYSA-N
General description
HX0770-6 1kg Poly Bottle
Application
Analysis Note
Assay: 96.0% min
Clarity of alcohol solution: To pass test
Color: White
Form: Crystals
Heavy metals (as Pb): 5ppm max
Iron (Fe): 5ppm max
Residue after ignition: 0.05% max
Sulfur compounds (as SO4): 0.005% max
Titrable free acid: 0.25meq/g max
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral
Target Organs
spleen
Storage Class Code
4.1A - Other explosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service