Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HX0200

Supelco

Hexadecane

Synonym(s):

Hexadecane, Cetane ASTM Grade

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$851.00

$851.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
250 G
$851.00

About This Item

Linear Formula:
CH3(CH2)14CH3
CAS Number:
MDL number:
UNSPSC Code:
12352001
EC Index Number:
208-878-9
vapor pressure:
<0.1 hPa ( 20 °C)

$851.00


In StockDetails


Request a Bulk Order

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

color

APHA: ≤10

mp

18 °C

transition temp

flash point 135 °C

shipped in

ambient

storage temp.

room temp

SMILES string

C(CCCCCCCC)CCCCCCC

InChI

1S/C16H34/c1-3-5-7-9-11-13-15-16-14-12-10-8-6-4-2/h3-16H2,1-2H3

InChI key

DCAYPVUWAIABOU-UHFFFAOYSA-N

Analysis Note

Assay (GC): 99.0% min
Color (APHA): 10 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

233.6 °F

Flash Point(C)

112 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service