Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HX0102

Sigma-Aldrich

1-Hexanesulfonic Acid, Sodium Salt, Monohydrate

HPLC

Synonym(s):

1-Hexanesulfonic Acid, Sodium Salt, Monohydrate, Hexane Sulfonic Acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$199.00
100 G
$803.00

$199.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$199.00
100 G
$803.00

About This Item

Linear Formula:
CH3(CH2)5SO3Na · H2O
CAS Number:
Colour Index Number:
73015
MDL number:
UNSPSC Code:
12352107
EC Index Number:
220-601-3

$199.00


Please contact Customer Service for Availability

Request a Bulk Order

mp

300 °C

shipped in

ambient

storage temp.

room temp

SMILES string

[Na+].[S](=O)(=O)([O-])CCCCCC.O

InChI

1S/C6H14O3S.Na.H2O/c1-2-3-4-5-6-10(7,8)9;;/h2-6H2,1H3,(H,7,8,9);;1H2/q;+1;/p-1

InChI key

CLCGFJYKZGFGSQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

1-Hexanesulfonic Acid, Sodium Salt, Monohydrate
HX0102-3 100g Poly Bottle

Application

  • Quantification of Riboflavin and Thiamine in GI Branded Yogurts Collected from Bogura, Bangladesh Using HPLC: This study utilized 1.5469 g of 1-hexanesulfonic acid sodium salt monohydrate in the HPLC fluorescence detection method to quantify vitamins in yogurt (Mia et al., 2022).
  • RP-HPLC method with indirect UV detection for determination of sodium ibandronate in pharmaceuticals: This study used 1-hexanesulfonic acid sodium salt as a mobile phase component for the RP-HPLC analysis of ibandronate in pharmaceuticals (Zharko et al., 2018).
  • OPTIMIZATION OF SEPARATION METHOD FOR THE ANALYSIS OF 5 PHARMACEUTICALS WITH ACETHYLCHOLINESTERASE INHIBITOR ACTIVITY: Describes the use of 1-hexanesulfonic acid sodium salt in the optimized separation method for analyzing pharmaceuticals with acetylcholinesterase inhibitor activity (Budi, 2015).

Analysis Note

Purity: 99% min
Solubility: To pass test
UV Abs. at 210 nm: 1.00AU max
UV Abs. at 220 nm: 0.10AU max
UV Abs. at 254 nm: 0.04AU max
UV Abs. at 280 nm: 0.04AU max

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service