Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HTS187RTA

Sigma-Aldrich

Ready-to-Assay Ghrelin Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$1,090.00

$1,090.00


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
$1,090.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.84

$1,090.00


Please contact Customer Service for Availability

Product Name

Ready-to-Assay Ghrelin Receptor Frozen Cells, Human Ghrelin / GHSR-1a / growth hormone secretagogue receptor GPCR frozen cells for Calcium Flux FLIPR Assays.

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

shipped in

dry ice

General description

Millipore’s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

Ghrelin is a 28 amino acid peptide, containing a unique octanoyl moiety added post-translationally to Ser3, with diverse activities in the CNS, gastrointestinal tract, and cardiovascular system (Davenport et al., 2005; Leite-Moreira and Soares, 2007). In the CNS, ghrelin stimulates release of growth hormone from the anterior pituitary and increases appetite by binding to neurons within the arcuate nucleus. Circulating concentrations of ghrelin increase with preprandially and decrease post-prandially, and thus counterbalances the effects of leptin to coordinate energy balance, appetite and food intake. Ghrelin is also expressed in the cardiovascular system where it acts as a potent vasodilator; receptors are up-regulated in patients with atherosclerosis, suggesting that it plays a role in compensating for increased vasoconstriction (Kleinz et al., 2006). The effects of ghrelin are mediated by a Gq-coupled receptor, originally designated GHSR (growth hormone secretogogue receptor), and more recently termed the Ghrelin Receptor or GRLN. Two splice variants have been described; type 1a (GHS-R1a) is the functional receptor, whereas type 1b (GHS-R1b) encodes a truncated, inactive protein with only 5 transmembrane domains. As administration of agonists of the ghrelin receptor to rats leads to increased food intake and antagonists reduce food intake (Beck et al. 2004), antagonism and inverse agonism of the ghrelin receptor may reduce food intake in certain types of obesity, and agonists of the ghrelin receptor are potentially useful for treatment of anorexia and cachexia. Millipore’s cloned human ghrelin-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant ghrelin expression on the cell surface for functional detection via the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at Ghrelin Receptor.

Cell Line Description

GPCR Cell Lines
Host cells: Chem-1

Biochem/physiol Actions

GPCR Class: A
Protein Target: Ghrelin / GHSR-1a / growth hormone secretagogue receptor
Target Sub-Family: Ghrelin

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.

Fifty (50) mL of Media Component.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service