Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HTS184RTA

Sigma-Aldrich

Ready-to-Assay CCK 1 Cholesystokinin Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76

product name

Ready-to-Assay CCK 1 Cholesystokinin Receptor Frozen Cells,

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

Cholecystokinins are a series of peptides of heterogeneous length (5 to 58 amino acids) that are derived from preprocholecystokinin and are found in gastrointestinal tissues and the central nervous system. Gastrin is a related peptide with 5 C-terminal amino acids identical to those of cholecystokinin. Two GPCRs, CCK1 (CCKA) and CCK2 (CCKB), bind to CCK and/or gastrin to mediate the biological effects of the peptides. CCK1 selectively binds sulfated CCK, whereas CCK2 binds to CCK and gastrin with similar affinity. Binding of ligands to CCK1 stimulates mobilization of intracellular calcium by activation of Gq/11. CCK1 receptors in the periphery are primarily localized in the pancreas, gallbladder, pylorus, and intestine where they are responsible of the regulation of diverse digestive processes. They are also present in select areas of the peripheral nervous system (vagus nerve), and the CNS where they mediate the satiety effects of CCK, regulate an increase in dopamine release, and antagonize opiod analgesia (Noble et al., 1999). Millipore′s cloned human CCK1-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant CCK1 expression on the cell surface for functional detection via the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists, and modulators at CCK1.

Cell Line Description

  • GPCR Cell Lines
  • Host cells: Chem-1

Application

Human CCK1 / CCKa GPCR frozen cells for Calcium Flux FLIPR Assays.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: CCK1/ CCKa
  • Target Sub-Family: Cholecystokinin

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.
Fifty (50) mL of Media Component.

Disclaimer


This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service