Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HTS125M

Sigma-Aldrich

ChemiScreen Membrane Preparation
Recombinant Human SST4 SOMATOSTATIN RECEPTOR

Human sst4 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.

Synonym(s):

ChemiScreen Receptor Kit, Human SST4 Membrane Prep, SST4 Somatostatin Receptor Prep

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.41

biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Human SSTR4 encoding SST4
Somatostatin is a 14 or 28 amino acid regulatory peptide that inhibits hormone secretion from the pituitary, pancreas, and other endocrine sites. A family of 6 GPCRs, sst1, sst2A, sst2B, sst3, sst4 and sst5, mediate the biological activity of somatostatins. The somatostatin receptors couple to Gi to inhibit cAMP production, and also increase MAP kinase signalling. Several tumors have been shown to overexpress somatostatin receptors, and binding of somatostatin to these tumor cells stimulates or inhibits proliferation, depending on the receptor subtypes expressed (Olias et al., 2004). Somatostatin has been implicated in seizure susceptibility in animal models, and activation of sst4 with selective agonists increases seizure activity (Moneta et al., 2002). Chemicon′s SST4 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of antagonists of SST4 interactions with somatostatin. The membrane preparations exhibit a Kd of 1.78 nM for [125I]-Somatostatin 14. With 5 ug/well SST4 Membrane Prep and 1 nM [125I]-Somatostatin 14, a greater than 20-fold signal-to-background ratio was obtained.

Application

Human sst4 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
Radioligand binding assay, and GTPγS binding.

Biochem/physiol Actions

GPCR Class: A
Protein Target: sst4
Target Sub-Family: Somatostatin

Packaging

200 units

Quality

SPECIFICATIONS: 1 unit = 5 μg membrane preparation
Bmax: 28.2 pmol/mg
Kd: 1.78 nM

Specifications

Inucbation Conditions
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50 mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C Radioligand: [125I]-Somatostatin 14 (Perkin Elmer # NEX389)
Wash Buffer: 50 mM Hepes, pH 7.4, 500 mM NaCl , 0.1% BSA, filtered and stored at 4°C.

Physical form

One package contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 20-fold signal:background with 125I-labeled somatostatin 14 at 1 nM. Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives. Packaging method: Membranes protein were adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation and function of somatostatin receptors
Olias, Gisela, et al
Journal of Neurochemistry, 89, 1057-1091 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service