Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HTS112RTA

Sigma-Aldrich

Ready-to-Assay Glucagon Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$1,070.00

$1,070.00


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
$1,070.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.84

$1,070.00


Please contact Customer Service for Availability

Product Name

Ready-to-Assay Glucagon Receptor Frozen Cells, Human glucagon / GCG GPCR frozen cells for Calcium flux Assays, ligand binding FLIPR Assays.

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable
ligand binding assay: suitable

shipped in

dry ice

General description

Millipore’s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

Glucagon is a 29-amino acid peptide that stimulates glycogenolysis and gluconeogenesis in the liver to increase blood glucose. The receptor for glucagon is a class 2 (or class B) GPCR that signals through Gs to stimulate cAMP production (Mayo et al., 2003). Mice lacking the glucagon receptor have mild hypoglycemia after fasting, and exhibit hyperplasia of pancreatic alpha-cells (Gelling et al., 2003). Because of its role in promoting hyperglycemia, the glucagon receptor presents a potential target for treatment of diabetes. Chemicon′s cloned human Glucagon Receptor-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant Glucagon Receptor expression on the cell surface and contains high levels of the promiscuous G protein Galpha15 to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for antagonists of interactions between the Glucagon Receptor and its ligands.

Cell Line Description

Host cells: Chem-1

Biochem/physiol Actions

Protein Target: glucagon / GCG

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongyan Shuai et al.
Acta physiologica (Oxford, England), 231(4), e13611-e13611 (2020-12-29)
cAMP typically signals downstream of Gs -coupled receptors and regulates numerous cell functions. In β-cells, cAMP amplifies Ca2+ -triggered exocytosis of insulin granules. Glucose-induced insulin secretion is associated with Ca2+ - and metabolism-dependent increases of the sub-plasma-membrane cAMP concentration ([cAMP]pm

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service