Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HTS110RTA

Sigma-Aldrich

Ready-to-Assay 5-HT 4B Serotonin Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76

product name

Ready-to-Assay 5-HT 4B Serotonin Receptor Frozen Cells,

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

5-Hydroxytryptamine (5-HT, also commonly known as serotonin) is synthesized in enterochromaffin cells in the intestine and in serotonergic nerve terminals. In the periphery, 5-HT mediates gastrointestinal motility, platelet aggregation, and contraction of blood vessels. Many functions of the central nervous system are influenced by 5-HT, including sleep, motor activity, sensory perception, arousal and appetite. A family of 12 GPCRs and one ion channel mediate the biological effects of 5-HT (Hoyer et al., 1994). 5-HT4 comprises at least 8 isoforms varying at the C-terminus, which are generated by alternative splicing. The expression and distribution of these splice variants differs among organs and tissues with many of them present in several tissues such as atrium, brain, and GI tract (Bockaert et al. 2004). To date, all isoforms have been shown to activate adenylyl cyclase in vitro, and no difference in signal transduction between C-terminal 5-HT4 receptor variants has been demonstrated. 5-HT4 receptors are of potential interest for the treatment of patients with GI motility disorders and Alzheimer′s disease (De Maeyer et al. 2008; Lezoualc′h 2007). Millipore′s cloned human 5HT4B-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant 5HT4B expression on the cell surface and contains high levels of the promiscuous G protein Gα15 to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at 5HT4B Receptor.

Cell Line Description

  • GPCR Cell Lines
  • Host cells: Chem-1

Application

Human 5-HT4 GPCR frozen cells for Calcium Flux FLIPR Assays.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: 5-HT4
  • Target Sub-Family: Serotonin

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.
Fifty (50) mL of Media Component.

Disclaimer


This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service