Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HTS064M

Sigma-Aldrich

ChemiSCREEN Human Angiotensin II Receptor, Type I Membrane Preparation

Human AT1 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84

biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

UniProt accession no.

shipped in

dry ice

Gene Information

human ... AGTR1(185)

General description

Angiotensin II (Ang II), an octapeptide produced by cleavage of angiotensinogen by angiotensin-converting enzyme, plays a fundamental role in cardiovascular homeostasis. Two GPCRs, AT1 and AT2, mediate the effects of AngII, although AT1 is primarily responsible for the effects of Ang II on renal function and development, thirst, electrolyte and water balance, and arterial blood pressure. Binding of Ang II to AT1 activates both Gq and Gi (De Gasparo et al., 2000). Chemicon′s AT1 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of antagonists of AT1 interactions and its ligands. The membrane preparations exhibit a Kd of 0.36 nM for [125I]-Sar1-Ile8-Angiotensin II. With 0.3 nM [125I]-Sar1-Ile8-Angiotensin II, 5 µg/well AT1 Membrane Prep yields greater than 50-fold signal-to-background ratio.
Full-length human cDNA encoding AT1

Application

Radioligand binding assay and GTPγS binding.

Biochem/physiol Actions

GPCR Class: A
Protein Target: AT1
Target Sub-Family: Angiotensin

Quality

Signal:background and specific binding values obtained in a competition binding assay with varying amounts of ATaub1 membrane prep:
tabletrth align="left"/tdth align="center"10 µg/well/tdth align="center"5 µg/well /td/trtrtd align="left" Signal:Background/tdtd align="center" 115.5/tdtd align="center" 92.8/td/trtrtd align="left" Specific Binding (cpm) /tdtd align="center"46,458/tdtd align="center"43,007 /td/tr/table

SPECIFICATIONS: 1 unit = 5 µg
Bmax for [125I] Angiotensin II binding: 5.53 pmol/mg protein;
Kd for [125I] Angiotensin II binding: ~ 0.36 nM

Specifications

Inucbation Conditions
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C
Radioligand: [125I]-Sar1-Ile8-Angiotensin II. (Perkin Elmer#:NEX-248 )
Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.
One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 50-fold signal:background with 125I labeled Sar1-Ile8-Angiotensin II at 0.3 nM

Physical form

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA no preservatives.
Packaging method: Membranes protein were adjusted to 0.5 mg/ml in 1 ml packaging buffer, rapidly frozen, and stored at -80oC.

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service