Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HTS062M

Sigma-Aldrich

ChemiScreen Recombinant Human NMU1 Neuromedin Receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.45

biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

form

liquid

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

General description

Human NMU1
Neuromedin U (NmU) is a peptide characterized by its ability to promote smooth muscle contraction (Brighton et al., 2004a). Two GPCRs, NMU1 and NMU2, mediate the contractile effects of neuromedin U by activation of both Gq and Gi (Brighton et al., 2004b). In addition, NmU binds to NMU1 expressed on Th2 cells to induce cytokine release (Johnson et al., 2004). Chemicon′s NMU1 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of antagonists of NMU1 interactions with neuromedin U25. The membrane preparations exhibit a Kd of 0.08 nM for [125I]-neuromedin U25. With 5 μg/well NMU1 Membrane Prep and 0.5 nM [125I]-neuromedin U25, a greater than 30-fold signal-to-background ratio was obtained.

Application

Human NMU1 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
Radioligand binding assay, and GTPγS binding.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: NMU1
  • Target Sub-Family: Neuromedin U

Specifications

Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.

Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C.

Radioligand: [125I]-neuromedin U25 (Perkin Elmer # NEX383)

Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.

Physical form

One package contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 30-fold signal:background with 125I-labeled neuromedin U25 at 0.5 nM.
Liquid in packaging buffer:

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service