Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HTS059M

Sigma-Aldrich

ChemiScreen V1A Vasopressin Membrane Preparation

Sign Into View Organizational & Contract Pricing

Select a Size

200 UNITS
$1,070.00

$1,070.00


Please contact Customer Service for Availability


Select a Size

Change View
200 UNITS
$1,070.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.45

$1,070.00


Please contact Customer Service for Availability

biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

form

liquid

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

General description

Arginine vasopressin (AVP) is a 9 amino acid peptide that functions as an antidiuretic, vasoconstrictor and neurotransmitter. The three vasopressin receptors, V1A, V1B and V2, are GPCRs; V1A and V1B couple to Gq and calcium release, whereas V2 couples to Gs. V1 receptors regulate fluid homeostasis by stimulating contraction of glomerular mesangial cells and renal vascular smooth muscle cells (Birnbaumer, 2000). Chemicon′s V1A membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of V1A interactions with its ligands. The membrane preparations exhibit a Kd of 55 pM for [125I]-vasopressin antagonist ([125I]-phenylacetyl-D-Tyr(Me)-Phe-Gln-Asn-Arg -Pro-Arg-Tyr-NH2). With 10 μg/well V1A Membrane Prep and 0.1 nM [125]-vasopressin V1A antagonist, greater than 12-fold signal-to-background ratio were obtained.
Human AVPR1 cDNA encoding V1A

Application

Human V1A GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
Radioligand binding assay, and GTPγS binding.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: V1A
  • Target Sub-Family: Vasopressin

Packaging

1 mL, 2 mg/mL

Physical form

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA no preservatives. Packaging method: Membranes protein was adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service