Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HTS047M

Sigma-Aldrich

ChemiScreen A1 Membrane Preparation

Human A1 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84

biological source

human

recombinant

expressed in Chem-3 cells

packaging

pkg of 1 mL

manufacturer/tradename

ChemiScreen
Chemicon®

concentration

0.5 mg/mL

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Extracellular adenosine mediates a multitude of biological effects, including wakefulness, antiarrythmia, bronchoconstriction and response to ischemia and oxidative stress. A family of four GPCR adenosine receptors, A1, A2A, A2B and A3, is responsible for these effects. The A1 receptor, which couples to Gi/o, is most highly expressed in brain, and mediates endogenous antinociception and neuronal response to hypoxia (Fredholm et al., 2001). A1 is also expressed in kidney, where it contributes to tubuloglomerular feedback. Chemicon′s cloned human A1-expressing cell line is made in the Chem-3 host, which supports high levels of recombinant A1 expression on the cell surface and contains high levels of the high levels of the promiscuous G protein Gα15 to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for antagonists of interactions between A1 and its ligands.

Chemicon′s A1 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of antagonists of A1 interactions with its ligands. The membrane preparations exhibit a Kd of 1.2 nM for [3H]-Cyclopentyl-1, 3-dipropylxanthine, 8-[dipropyl-2, 3-3H (N)]. With 5mg/well A1 Membrane Prep and 20nM [3H]-DPCPX, a greater than 6-fold signal-to-background ratio is obtained.

Biochem/physiol Actions

Protein Target: A1
Target Sub-Family: Adenosine

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service