Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HTS025RTA

Sigma-Aldrich

Ready-to-Assay MCH 1 Melanin-Concentrating Hormone Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76

product name

Ready-to-Assay MCH 1 Melanin-Concentrating Hormone Receptor Frozen Cells,

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

Melanin-concentrating hormone (MCH) is a cyclic, 19 amino acid peptide produced in the lateral hypothalamus and other regions of the mammalian brain (Bittencourt et al., 1992). When administered intracerebrally or overexpressed transgenically in rodents, MCH stimulates food intake and weight gain (Ludwig et al., 2001). A receptor for MCH, previously identified as an orphan receptor SLC-1 and renamed MCH1, has been identified in humans and rodents. Consistent with a role in hypothalamic control of feeding behavior, MCH1 is expressed in in the ventromedial and dorsomedial nuclei of the hypothalamus (Chambers et al., 1999). Genetic ablation and pharmacological antagonism of MCH1 in rodents results in resistance to obesity caused by a high fat diet or leptin deficiency (Borowsky et al., 2002; Segal-Lieberman, et al., 2003; Shearman et al., 2003; Marsh et al., 2002; Chen et al., 2002). Thus, MCH1 is an attractive target for obesity. Millipore′s cloned human MCH1-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant MCH1 expression on the cell surface for functional detection via the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists, and modulators at MCH1.

Cell Line Description

  • GPCR Cell Lines
  • Host cells: Chem-1

Application

Human MCHR1 / GPR24 GPCR frozen cells for Calcium Flux FLIPR Assays.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: MCHR1/ GPR24
  • Target Sub-Family: Melanin-concentrating hormone

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.
Fifty (50) mL of Media Component.

Disclaimer


This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service