Skip to Content
MilliporeSigma
All Photos(1)

Documents

HTS017RTA

Sigma-Aldrich

Ready-to-Assay C5a Anaphylotoxin Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76

product name

Ready-to-Assay C5a Anaphylotoxin Receptor Frozen Cells,

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

C5a is a proinflammatory peptide generated through activation of the complement system, and is more potent than the other anaphylatoxins, C4a and C3a, in activating peripheral blood leukocytes (Gerard and Gerard, 1994). It can actively participate in the regulation of local cytokine network by stimulating production of proinflammatory cytokines (Buchner et al., 1995; Hsu et al., 1999). The proinflammatory effects of C5a are mediated through binding to a specific 7-TM chemoattractant receptor, C5aR (CD88). Binding of C5a to phagocyte C5aR induces chemotaxis, production of superoxide anions, and release of degradative enzymes. Pharmacologic or genetic disruption of C5a/C5aR interaction reduces sepsis (Riedemann et al., 2003), immune complex-induced lung disease (Shushakova et al., 2002), and Arthrogen-induced arthritis (Grant et al., 2002) in experimental models. Millipore′s cloned human C5a-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant C5a expression on the cell surface and contains high levels of the promiscuous G protein Gα15 to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists, and modulators at C5a.

Cell Line Description

  • GPCR Cell Lines
  • Host cells: Chem-1

Application

Human C5aR GPCR frozen cells for Calcium Flux FLIPR Assays.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: C5aR
  • Target Sub-Family: Anaphylotoxin

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.
Fifty (50) mL of Media Component.

Disclaimer


This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service