Skip to Content
MilliporeSigma
All Photos(1)

Documents

HTS016M

Sigma-Aldrich

ChemiSCREEN C3aR Membrane Preparation

Human C3aR GPCR membrane preparation for Radioligand binding Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84

biological source

human

Quality Level

recombinant

expressed in Chem-2 cells

manufacturer/tradename

ChemiScreen
Chemicon®

concentration

0.5 mg/mL

technique(s)

radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

C3a, along with C4a and C5a, is a 77 amino acid anaphylotoxin generated by proteolytic cleavage during activation of the complement pathway (Ember, 1997). The anaphylotoxins strongly promote inflammation by recruiting leukocytes, particularly basophils, eosinophils, neutrophils and monocyte (Martin, 1997). The proinflammatory properties of C3a are mediated by interaction between the peptide and a 7-TM G-protein coupled receptor, C3aR (Crass, 1996). Genetic and pharmacological inhibition of C3a/C3aR interaction indicates an important role for C3aR in allergic asthsma Humbles, 2000; Drouin, 2002). C3a/C3aR also enhances the effect of SDF-1 in promoting retention of haematopoietic stem/progenitor cells within the bone marrow (Rataiczak, 2004). Chemicon′s C3aR membrane preps are an ideal tool for screening for compounds that disrupt binding of C3a to C3aR. The membranes have been shown to bind to 125I-labeled C3a with a Kd of 0.06 nM.
Full-length human C3aR cDNA

Application

Human C3aR GPCR membrane preparation for Radioligand binding Assays.
Radioligand binding assay

Biochem/physiol Actions

GPCR Class: A
Protein Target: C3aR
Target Sub-Family: Anaphylotoxin

Quality

Bmax for [125I]C3a binding: 1.9 pmol/mg protein
Kd for [125I] C3a binding: ~ 0.06 nM

Physical form

Membranes were adjusted to 0.5mg protein/mL in 50 mM Tris-HCL, pH 7.4, 10% glycerol and 1% BSA, rapidly frozen, and stored at -80°C. One vial contains 500 units, where a unit is the amount of membrane that will yield greater than 1000 cpm specific binding of 125I-labeled C3a (0.04 nM input concentration).

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service