Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HTS004M

Sigma-Aldrich

ChemiSCREEN CXCR4 Membrane Preparation

Human CXCR4 GPCR membrane preparation for Radioligand binding Assays & GTPgammaS binding.

Sign Into View Organizational & Contract Pricing

Select a Size

200 UNITS
$1,050.00

$1,050.00


Please contact Customer Service for Availability


Select a Size

Change View
200 UNITS
$1,050.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84

$1,050.00


Please contact Customer Service for Availability

biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

packaging

pkg of 2 mL

manufacturer/tradename

ChemiScreen
Chemicon®

concentration

0.5 mg/mL

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... CXCR4(7852)

General description

Full-length human CXCR4 cDNA
The chemokine SDF-1a and its GPCR receptor CXCR4 have a one-to-one specificity that is unique among chemokines and their receptors. SDF-1a binds to CXCR4 expressed on hematopoietic and lymphopoietic cells, and directs their trafficking to and retention in hemato-and lymphatopoietic organs and sites of inflammation (Kucia et al., 2004). CXCR4 is expressed on several tumor cell lineages, and might be responsible for metastasis to sites of SDF-1a expression, such as bone and lymph nodes (Muller et al., 2001). In addition, CXCR4 is a coreceptor for the HIV envelope glycoprotein gp120 (Feng et al., 1996). Small molecule antagonists of CXCR4 have been developed and shown to inhibit infectivity of T-tropic HIV strains and to impair growth of brain tumors (Arakaki et al., 1999; Rubin et al., 2003). Chemicon′s CXCR4 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal tools for screening for antagonists of interactions between CXCR4 and SDF-1a. Radiolabeled SDF-1a binds to the CXCR4 membranes with a Kd of 0.9 nM. With 0.04-0.13 nM 125I-labeled SDF-1a, an approximately 3-fold window of specific binding is obtained.

Application

Radioligand binding assay and GTPgammaS binding.

Biochem/physiol Actions

Protein Target: CXCR4

Quality



Table 1. Signal:background and specific binding values obtained in a competition binding assay with CXCR4 membrane prep, 0.13 nM 125I-SDF-1α and a dilution series of unlabeled SDF-1α.
5 µg/well
Signal:Background 4.9
Specific Binding (cpm) 9004


Specifications:
1 unit = 5 μg
Bmax [125I]-SDF-1alpha binding: 1.6 pmol/mg protein
Kd for [125I] SDF-1alpha binding: ~ 0.9 nM

Specifications

Inucbation Conditions
RECOMMENDED ASSAY CONDITIONS: Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50 mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.

Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C

Radioligand: [125I] SDF-1alpha (PerkinElmer#: NEX346)

Wash Buffer: 50 mM Hepes, pH 7.4, 500 mM NaCl , 0.1% BSA, filtered and stored at 4°C.

One vial contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 4-fold signal:background with 125I-labeled SDF-1alpha at 0.13 nM.

Physical form

Packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA

Packaging method: Membranes protein were adjusted to 0.5 mg/ml in 1 ml packaging buffer, rapidly frozen, and stored at -80°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Feng et al.
Science (New York, N.Y.), 272(5263), 872-877 (1996-05-10)
A cofactor for HIV-1 (human immunodeficiency virus-type 1) fusion and entry was identified with the use of a novel functional complementary DNA (cDNA) cloning strategy. This protein, designated "fusin," is a putative G protein-coupled receptor with seven transmembrane segments. Recombinant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service