Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HG000C98

Millipore

Chloramphenicol Agar

pH 6.6±0.2 (25 °C, after autoclaving)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352111

composition

Agar, 12.0 g/L
Chloramphenicol, 0.1 g/L
Dextrose, 20.0 g/L
Yeast Extract, 5.0 g/L

packaging

poly bottle of 12 × 500 g (HG000C98.500)

pH

6.6±0.2 (25 °C, after autoclaving)

storage temp.

15-25°C

Application

A selective medium suitable for the detection of yeasts and moulds in food.

Preparation Note

Suspend 37g in 1 litre demineralised water. Boil whilst stirring until completely dissolved. Autoclave at 121°C for 15 minutes. Cool agar to 45 - 50°C, mix well and pour plates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service