Skip to Content
MilliporeSigma
All Photos(1)

Documents

HG000C92

Millipore

m-FC Agar

pH 7.4±0.2 (25 °C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352111

composition

Agar, 13.0 g/L
Aniline Blue, 0.1 g/L
Bile Salts No 3, 1.5 g/L
Lactose, 12.5 g/L
Peptone, 5.0 g/L
Sodium chloride, 5.0 g/L
Tryptone, 10.0 g/L
Yeast Extract, 3.0 g/L

packaging

poly bottle of 12 × 500 g (HG000C92.500)

pH

7.4±0.2 (25 °C)

storage temp.

15-25°C

Application

An agar suitable for the detection and enumeration of faecal coliforms by the membrane filtration technique.

Preparation Note

Suspend 50g in 1 litre demineralised water. Boil whilst stirring until completely dissolved. If desired, add 10ml of a 1% solution of rosolic acid in 0.2N NaOH to the medium. Continue heating for 1 minute whilst stirring. Cool to 45 -50°C. Mix well and pour plates.

Do not Autoclave or Overheat

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service