Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HG000C74

Millipore

m-Endo Agar Les

pH 7.2±0.2 (25 °C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352111

composition

Agar, 15.0 g/L
Basic Fuchsin, 0.8 g/L
di-Potassium Hydrogen Phosphate, 3.3 g/L
Lactose, 9.4 g/L
Peptone, 3.7 g/L
Potassium Dihydrogen Phosphate, 1.0 g/L
Sodium Chloride, 3.7 g/L
Sodium Deoxycholate, 0.1 g/L
Sodium Lauryl Sulphate, 0.05 g/L
Sodium Sulphate, 1.6 g/L
Tryptone, 3.7 g/L
Tryptose, 7.5 g/L
Yeast Extract, 1.2 g/L

packaging

poly bottle of 12 × 500 g (HG000C74.500)

pH

7.2±0.2 (25 °C)

storage temp.

15-25°C

Application

A selective culture medium for the enumeration of coliforms in water, using a membrane filter method.

Preparation Note

To ensure complete solution of basic fuchsin, prepare medium as follows: Suspend 51g in 1 litre demineralised water. Add 20ml ethanol and stand for 10 minutes. Boil until agar is dissolved. Pour into plates. Store away from light at 4°C.

Do not Autoclave or Overheat

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service