Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HG000C21

Millipore

XLD Agar

pH 7.4±0.2 (25 °C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352111

composition

Agar, 10.5 g/L
Ferric Ammonium Citrate, 0.8 g/L
Lactose, 7.5 g/L
L(+) Lysine, 5.0 g/L
Phenol Red, 0.08 g/L
Sodium chloride, 5.0 g/L
Sodium Deoxycholate, 2.5 g/L
Sodium Thiosulphate, 6.8 g/L
Sucrose, 7.5 g/L
D(+) Xylose, 3.5 g/L
Yeast Extract, 3.0 g/L

packaging

poly bottle of 12 × 500 g (HG000C21.500)

pH

7.4±0.2 (25 °C)

storage temp.

15-25°C

Application

A medium suitable for the isolation, cultivation and differentiation of pathogenic Enterobacteriaceae especially Shigella and Salmonella species.

Preparation Note

Suspend 52g in 1 litre demineralised water. Boil whilst stirring until completely dissolved. Cool to 45 - 50°C, mix well and pour plates.

Do not Autoclave or Overheat

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service