Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HG000BX1

Millipore

Agar Bacteriological

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

packaging

pkg of 25 kg (HG000BX1.25)
pkg of 12 × 500 g (HG000BX1.500, plastic bottles)

storage temp.

2-30°C

Application

Biolab Agar Bacteriological is a special grade of agar, purified and tested for general microbiological use. The recommended amount used in solid media is 12g per litre or up to 15g if liquids such as blood are to be incorporated. 0.05 - 0.2% agar added to liquid media will promote growth of anaerobes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service