Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HG0000C5

Millipore

Sabouraud Dextrose 4% Agar

pH 5.6±0.2 (25 °C, after autoclaving)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352111

composition

Agar, 10.0 g/L
Dextrose, 40.0 g/L
Peptone, 10.0 g/L

packaging

poly bottle of 12 × 500 g

pH

5.6±0.2 (25 °C, after autoclaving)

storage temp.

15-25°C

Application

The medium is used in the isolation, cultivation and identification of yeasts and fungi and is particularly useful in the differentiation of dermatophytes.

Preparation Note

Suspend 60g in 1 litre demineralised water. Boil whilst stirring until completely dissolved. Autoclave at 121°C for 15 minutes. Cool agar to 45 - 50°C, mix well and pour plates.
500mg Cycloheximide and 40mg Chloramphenicol may be added to each litre of medium before autoclaving to inhibit non-pathogenic fungi or bacteria.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service