Hi-Flow Plus membranes are not characterized by pore size. However, the HF180 membrane is estimated to have a pore size between 5 microns and 8 microns, based on expected performance in traditional filtration methods.
HF180MC100
Hi-Flow™ Plus Membrane 180
pack of 100 laminated membrane cards, size 6 cm × 30.1 cm
Synonym(s):
HF, Hi-Flow, Hi-Flow™ Plus HF180 Membrane Cards, immunoassay membrane, lateral flow membrane, membrane card, nitrocellulose membrane
Select a Size
Select a Size
About This Item
packaging
pack of 100 laminated membrane cards
Quality Level
parameter
180 sec/4 cm capillary flow rate
size
6 cm × 30.1 cm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Hi-Flow™ Plus membrane is a critical raw material suitable for use in a broad range of Lateral Flow test applications, including Infectious Disease testing (COVID-19, HIV, Influenza, Malaria, STDs etc.), as well as in women’s health, biomarker detection, drugs of abuse testing, drug development, environmental testing, agricultural applications, food safety, product quality testing and animal health.
- Hi-Flow™ Plus membranes are suitable for use with all lateral flow test techniques including competitive, sandwich, and multiplex detection assays.
- They are optimised for use in both qualitative and quantitative test types and pair well with lateral flow detector particles including colloidal gold nanoparticles, dyed/ fluorescent microspheres etc.
- Hi-Flow™ Plus membranes are suitable for use in lateral flow tests designed to detect analytes present in a broad range of samples matrices including, blood, saliva, urine, nasopharyngeal swabs, sweat and others.
Features and Benefits
- Consistent Flow & Surface Chemistry, Robust manufacturing process
- Proprietary Surface Chemistry Optimised for Lateral Flow Applications
- High Quality Level: MQ400
- Trusted Partner and Brand with Decades of membrane manufacturing experience
- High production capacity, high stock levels, multiple manufacturing lines
- Convenient samples packs available in several formats
- Manufacturing scale quantities available to purchase online with immediate shipping.
- Consistent capillary flow time and membrane thickness
- Superior surface morphology allows for consistent and even flow distribution of reagents across the membrane surface
- Sharp, intense, and uniform test and control line formation
- Excellent intra-lot and inter-lot consistency.
- Better Assay sensitivity
- Excellent reproducibilityImproved lateral flow test manufacturing yields.
- Quick Delivery and Lead time.
- No supply disruption
- Excellent Technical support
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the pore size of the nitrocellulose membrane?
1 answer-
Helpful?
-
-
are there plastic boxes that hold the membrane to assemble the detection devices, including single-plex and double-plex holders?
1 answer-
Regrettably, we do not offer injection molded or 3D printed cartridge options for the final assembly of a device.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service