Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HCYTAAB-17K

Millipore

MILLIPLEX® Human Cytokine Autoantibody Panel

Sign Into View Organizational & Contract Pricing


About This Item

NACRES:
NA.47

To order a Milliplex® kit, please search for your analyte of interest.

species reactivity

human

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

Anti-cytokine antibodies occur frequently and are present in healthy individuals and patients with acquired immunodeficiency and autoimmune diseases. Cytokines offering protection against microbes can be targeted by cytokine autoantibodies, leading to life-threatening infections. Some infections associated with anti-cytokine antibodies include M. tuberculosis, Salmonella, S. aureus. E. coli, Histoplasma, and more. The presence of cytokine autoantibodies in autoimmune disease can influence disease severity and activity. These autoimmune diseases may include systemic lupus erythematosus (SLE), multiple sclerosis (MS), Sjögren′s syndrome, rheumatoid arthritis (RA), and acute respiratory distress syndrome. Measuring cytokine autoantibodies may be useful for disease monitoring and efficacy of treatment.The MILLIPLEX® MAP Human Cytokine Autoantibody Magnetic Bead Panel is a 15-plex kit to be used for the simultaneous detection and profiling of any or all of the following autoantibody analytes in serum and plasma samples: Anti-BAFF, Anti-G-CSF, Anti-IFNβ, Anti-IFNγ, Anti-IL-1α, Anti-IL-6, Anti-IL-8, Anti-IL-10, Anti-IL-12 (p40), Anti-IL-15, Anti-IL-17A, Anti-IL-17F, Anti-IL-18, Anti-IL-22, and Anti-TNFα. For this particular panel, 4 assay control beads are also included as part of the base kit format.

Application

The Human Cytokine Autoantibody panel is a bead-based multiplex assay based on the Luminex® xMAP technology which enables the simultaneous analysis of cytokine autoantibodies in human serum and plasma samples.This is assay yields results in Median Fluorescence Intensity (MFI).Four assay control beads are included in the base kit.This is an overnight or 2 hour incubation assay.This assay requires 25 μL of 1:100 diluted plasma or serum.Analytes available:BAFF/Blys;G-CSF;IFNβ;IFNγ;IL-1α;IL-6;IL-8;IL-10;IL-12 (p40);IL-15;IL-17A;IL-17F;IL-18;IL-22;TNFα

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

96-well plate

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Multiplex analysis of autoantibodies, such as MILLIPLEX® multiplex assays, helps advance research by detecting multiple autoantibodies like cytokine autoantibodies, autoimmune autoantibodies, and cancer autoantibodies at once to give researchers a full picture of analyte concentrations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service