The HC5B-140K single plex has been tested with HCMPEX1-19K and HCMP2MAG-19K. Cross reactivity is observed with C5, C5a, and C9 in HCMPEX1-19K. However, no cross reactivity is observed with any analytes in HCMP2MAG-19K. There hasn't been testing for cross reactivity between HCMPEX1-19K and HCMP2MAG-19K.
HCMPEX1-19K
MILLIPLEX® Human Complement Expanded Panel 1
About This Item
Recommended Products
manufacturer/tradename
Milliplex®
Quality Level
technique(s)
multiplexing: suitable
detection method
fluorometric
shipped in
wet ice
storage temp.
2-8°C
General description
The complement system consists of three different simultaneous pathways. The classical pathway is stimulated by antigen-antibody complexes; the alternative pathway spontaneously activates on contact with pathogenic cell surfaces; the mannose-binding lectin pathway recognizes mannose sugars usually present only on pathogenic cell surfaces.
The complement system is thought to play a key role in many diseases with an immune component, such as asthma or sepsis, and in many autoimmune diseases, including systemic lupus erythematosus, inflammatory bowel disease, rheumatoid arthritis, and multiple sclerosis. It is also becoming increasingly associated with neurological diseases, such as Alzheimer’s Disease, and conditions such as spinal cord injuries. The MILLIPLEX® Human Complement Expanded Panel 1 is a 9-plex kit to be used for the simultaneous quantification of any or all of the following analytes in serum, plasma, or culture supernatant samples: C2, C4b, C5, C5a, C9, Factor D (Adipsin), Factor I, Mannose-Binding Lectin (MBL), and Properdin.
Packaging
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2
Target Organs
Respiratory Tract
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Immunology multiplex assays, like MILLIPLEX® assays, allow researchers to investigate more immune biomarkers simultaneously, saving time and resources.
-
Is there any available data on mixing and matching the complement markers across the three panels? Additionally, is there any data on spiking the C5b-9 analyte into the other two panels?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service